(5-Brompicolinoyl)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-(piperidin-1-ylcarbonyl)pyridine, (5-Bromopyridin-2-yl)(piperidin-1-yl)methanone

PY-7213-1G PY-7213-5G
COBBPY-7213-1GEA 251 EUR
COBBPY-7213-1G COBBPY-7213-5G
(5-Brompicolinoyl)piperidin
(5-Brompicolinoyl)piperidin
Formel: C₁₁H₁₃BrN₂O
Molecular Weight: 269.1377 g/mol
MDL: MFCD08458472
CAS-Nummer: 934000-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR