N(α)-Fmoc-L-tryptophan
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Fmoc-Trp-OH
4003185.0025
4003185.0100
4003185.0500
BACE4003185.0025EA
0
EUR
BACE4003185.0025
BACE4003185.0100
BACE4003185.0500
N(α)-Fmoc-L-tryptophan
N(α)-Fmoc-L-tryptophan
Formel:
C₂₆H₂₂N₂O₄ Molecular Weight: 426,5 g/mol |
MDL:
MFCD00037126 CAS-Nummer: 35737-15-6 EINECS: 252-706-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...