Ethyl(tetrahydropyran-4-yliden)acetat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-(dihydro-2H-pyran-4(3H)-ylidene)acetate

AM-1009-5G AM-1009-1G
COBBAM-1009-5GEA 127 EUR
COBBAM-1009-5G COBBAM-1009-1G
Ethyl(tetrahydropyran-4-yliden)acetat
Ethyl(tetrahydropyran-4-yliden)acetat
Formel: C₉H₁₄O₃
Molecular Weight: 170,2057 g/mol
MDL: MFCD09475810
CAS-Nummer: 130312-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR