2-(Morpholinomethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine
PN-5067-5G
PN-5067-1G
COBBPN-5067-5GEA
0
EUR
COBBPN-5067-5G
COBBPN-5067-1G
2-(Morpholinomethyl)phenylboronsäurepinakolester
2-(Morpholinomethyl)phenylboronsäurepinakolester
Formel:
C₁₇H₂₆BNO₃ Molecular Weight: 303.2043 g/mol |
MDL:
MFCD07781208 CAS-Nummer: 876316-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...