4-(Morpholinomethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester, 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 4,4,5,5-Tetramethyl-2-[4-(morpholinomethyl)phenyl]-1,3,2-dioxaborolane, 4-(Morpholinomethyl)phenylboronic acid Pinacol ester

PN-8855-1G PN-8855-5G
COBBPN-8855-1GEA 50.8 EUR
COBBPN-8855-1G COBBPN-8855-5G
4-(Morpholinomethyl)phenylboronsäurepinakolester
4-(Morpholinomethyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₃
Molecular Weight: 303,20 g/mol
MDL: MFCD04974052
CAS-Nummer: 364794-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR