3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Morpholino(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone, 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester

PN-3054-1G PN-3054-5G
COBBPN-3054-1GEA 48.4 EUR
COBBPN-3054-1G COBBPN-3054-5G
3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester
3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₄BNO₄
Molecular Weight: 317.19 g/mol
MDL: MFCD09266172
CAS-Nummer: 1036991-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR