3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Morpholino(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone, 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
M-Clarity™ Qualitätsstufe = MQ100
699438-1G
699438-5G
SIAL699438-1GEA
44.2
EUR
SIAL699438-1G
SIAL699438-5G
3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester
Formel:
C₁₇H₂₄BNO₄ Molecular Weight: 317,19 g/mol |
MDL:
MFCD09266172 CAS-Nummer: 1036991-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...