4-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester, 4-(Morpholine-4-carbonyl)phenylboronic Acid Pinacol Ester

M-Clarity™ Qualitätsstufe = MQ100

677809-1G 677809-5G
SIAL677809-1GEA 56.2 EUR
SIAL677809-1G SIAL677809-5G
4-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(4-Morpholinylcarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₄BNO₄
Molecular Weight: 317,19 g/mol
MDL: MFCD05863913
CAS-Nummer: 656239-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR