2-(Morpholinomethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-(Morpholinomethyl)phenyl)boronic acid
BB-5067-1G
BB-5067-5G
COBBBB-5067-1GEA
0
EUR
COBBBB-5067-1G
COBBBB-5067-5G
2-(Morpholinomethyl)phenylboronsäure
2-(Morpholinomethyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₃ Molecular Weight: 221.0600 g/mol |
MDL:
MFCD01319041 CAS-Nummer: 223433-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...