3-(Morpholinomethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-(Morpholinomethyl)phenyl)boronic acid
BB-5196-5G
BB-5196-1G
COBBBB-5196-5GEA
0
EUR
COBBBB-5196-5G
COBBBB-5196-1G
3-(Morpholinomethyl)phenylboronsäure
3-(Morpholinomethyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₃ Molecular Weight: 221.0600 g/mol |
MDL:
MFCD08060623 CAS-Nummer: 397843-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...