2-(4-Morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2954-5G BB-2954-1G
COBBBB-2954-5GEA 2070 EUR
COBBBB-2954-5G COBBBB-2954-1G
2-(4-Morpholinylcarbonyl)phenylboronsäure
2-(4-Morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₄
MDL: MFCD08235036
CAS-Nummer: 874219-17-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR