3-(4-Morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(morpholine-4-carbonyl)phenyl)boronic acid, 3-(Morpholine-4-carbonyl)phenylboronic acid, 3-(4-Morpholinylcarbonyl)benzeneboronic acid

BB-3054-5G BB-3054-1G
COBBBB-3054-5GEA 931 EUR
COBBBB-3054-5G COBBBB-3054-1G
3-(4-Morpholinylcarbonyl)phenylboronsäure
3-(4-Morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₄
Molecular Weight: 235.05 g/mol
MDL: MFCD04115687
CAS-Nummer: 723281-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR