5-Chlor-2-(4-morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5236-5G BB-5236-1G
COBBBB-5236-5GEA 884 EUR
COBBBB-5236-5G COBBBB-5236-1G
5-Chlor-2-(4-morpholinylcarbonyl)phenylboronsäure
5-Chlor-2-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BClNO₄
MDL: MFCD11053808
CAS-Nummer: 1072946-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR