4-Chlor-3-(4-morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-3-(4-morpholinylcarbonyl)benzeneboronic acid, (4-Chloro-3-(morpholine-4-carbonyl)phenyl)boronic acid

BB-2815-5G BB-2815-1G
COBBBB-2815-5GEA 1580 EUR
COBBBB-2815-5G COBBBB-2815-1G
4-Chlor-3-(4-morpholinylcarbonyl)phenylboronsäure
4-Chlor-3-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BClNO₄
Molecular Weight: 269.49 g/mol
MDL: MFCD07783863
CAS-Nummer: 871332-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR