3-Chlor-5-(4-morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-5-(morpholine-4-carbonyl)phenylboronic acid

BB-3244-5G BB-3244-1G
COBBBB-3244-5GEA 1310 EUR
COBBBB-3244-5G COBBBB-3244-1G
3-Chlor-5-(4-morpholinylcarbonyl)phenylboronsäure
3-Chlor-5-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BClNO₄
MDL: MFCD09800860
CAS-Nummer: 957120-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR