5-Fluor-2-(4-morpholinylcarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-6019-1G
BB-6019-250M
COBBBB-6019-1GEA
1180
EUR
COBBBB-6019-1G
COBBBB-6019-250M
5-Fluor-2-(4-morpholinylcarbonyl)phenylboronsäure
5-Fluor-2-(4-morpholinylcarbonyl)phenylboronsäure
Formel:
C₁₁H₁₃BFNO₄ |
MDL:
MFCD12913944 CAS-Nummer: 1217501-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...