4-Fluor-3-(4-morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-3-(morpholine-4-carbonyl)phenylboronic acid

BB-2872-1G BB-2872-5G BB-2872-250M
COBBBB-2872-1GEA 0 EUR
COBBBB-2872-1G COBBBB-2872-5G COBBBB-2872-250M
4-Fluor-3-(4-morpholinylcarbonyl)phenylboronsäure
4-Fluor-3-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₄
MDL: MFCD08235048
CAS-Nummer: 874219-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR