3-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Fluoro-5-(morpholine-4-carbonyl)phenylboronic acid, 3-Fluoro-5-(4-morpholinylcarbonyl)benzeneboronic acid
BB-2856-1G
BB-2856-5G
COBBBB-2856-1GEA
0
EUR
COBBBB-2856-1G
COBBBB-2856-5G
3-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure
3-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure
Formel:
C₁₁H₁₃BFNO₄ Molecular Weight: 253.04 g/mol |
MDL:
MFCD08235057 CAS-Nummer: 874219-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...