2-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-5-(morpholine-4-carbonyl)phenylboronic acid, (2-Fluoro-5-(morpholine-4-carbonyl)phenyl)boronic acid, 2-Fluoro-5-(4-morpholinylcarbonyl)benzeneboronic acid

BB-5300-1G BB-5300-5G BB-5300-250M
COBBBB-5300-1GEA 548 EUR
COBBBB-5300-1G COBBBB-5300-5G COBBBB-5300-250M
2-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure
2-Fluor-5-(4-morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₃BFNO₄
Molecular Weight: 253.04 g/mol
MDL: MFCD11504847
CAS-Nummer: 1072951-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR