3-(4-Thiomorpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Thiomorpholine-4-carbonyl)phenyl)boronic acid

BB-3091-5G BB-3091-1G
COBBBB-3091-5GEA 1580 EUR
COBBBB-3091-5G COBBBB-3091-1G
3-(4-Thiomorpholinylcarbonyl)phenylboronsäure
3-(4-Thiomorpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₃S
Molecular Weight: 251,1100 g/mol
MDL: MFCD06659887
CAS-Nummer: 850567-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR