4-(4-Thiomorpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Thiomorpholine-4-carbonyl)phenylboronic acid, (4-(Thiomorpholine-4-carbonyl)phenyl)boronic acid

BB-3041-5G BB-3041-1G
COBBBB-3041-5GEA 1580 EUR
COBBBB-3041-5G COBBBB-3041-1G
4-(4-Thiomorpholinylcarbonyl)phenylboronsäure
4-(4-Thiomorpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₃S
Molecular Weight: 251,1100 g/mol
MDL: MFCD06659879
CAS-Nummer: 850568-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR