4-(1-Piperazinylcarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Piperazin-1-yl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

PN-8872-1G PN-8872-250M
COBBPN-8872-1GEA 246 EUR
COBBPN-8872-1G COBBPN-8872-250M
4-(1-Piperazinylcarbonyl)phenylboronsäurepinakolester
4-(1-Piperazinylcarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₅BN₂O₃
Molecular Weight: 316,2030 g/mol
MDL: MFCD06657704
CAS-Nummer: 864754-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR