4-(4-Boc-1-piperazinylcarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperazine-1-carboxylate

PN-8909-1G PN-8909-5G
COBBPN-8909-1GEA 0 EUR
COBBPN-8909-1G COBBPN-8909-5G
4-(4-Boc-1-piperazinylcarbonyl)phenylboronsäurepinakolester
4-(4-Boc-1-piperazinylcarbonyl)phenylboronsäurepinakolester
Formel: C₂₂H₃₃BN₂O₅
MDL: MFCD08706021
CAS-Nummer: 864754-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR