4-(4-Boc-1-piperazinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperazine-1-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

684287-1G 684287-5G
SIAL684287-1GEA 94.5 EUR
SIAL684287-1G SIAL684287-5G
4-(4-Boc-1-piperazinylcarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(4-Boc-1-piperazinylcarbonyl)phenylboronsäurepinakolester
Formel: C₂₂H₃₃BN₂O₅
Molecular Weight: 416,32 g/mol
MDL: MFCD08706021
CAS-Nummer: 864754-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR