3-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-3846-1G BB-3846-5G
COBBBB-3846-1GEA 364 EUR
COBBBB-3846-1G COBBBB-3846-5G
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid
Formel: C₁₂H₁₈BClN₂O₃
MDL: MFCD09027254
CAS-Nummer: 957060-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR