3-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-3845-5G BB-3845-1G
COBBBB-3845-5GEA 0 EUR
COBBBB-3845-5G COBBBB-3845-1G
3-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid
3-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid
Formel: C₁₃H₂₀BClN₂O₄
MDL: MFCD09027253
CAS-Nummer: 957060-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR