4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid ≥98%
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
(4-(4-(2-Hydroxyethyl)piperazine-1-carbonyl)phenyl)boronic acid hydrochloride
BB-3214-1G
BB-3214-5G
COBBBB-3214-1GEA
0
EUR
COBBBB-3214-1G
COBBBB-3214-5G
4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid ≥98%
4-(4-(2-Hydroxyethyl)-1-piperazinylcarbonyl)phenylboronsäure Hydrochlorid
Formel:
C₁₃H₁₉BN₂O₄·HCl Molecular Weight: 314.57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027218 CAS-Nummer: 913835-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...