3-(4-Boc-1-piperazinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(4-(tert-Butoxycarbonyl)piperazine-1-carbonyl)phenyl)boronic acid

BB-8802-5G BB-8802-1G
COBBBB-8802-5GEA 0 EUR
COBBBB-8802-5G COBBBB-8802-1G
3-(4-Boc-1-piperazinylcarbonyl)phenylboronsäure
3-(4-Boc-1-piperazinylcarbonyl)phenylboronsäure
Formel: C₁₆H₂₃BN₂O₅
Molecular Weight: 334.175 g/mol
MDL: MFCD12546575
CAS-Nummer: 1218790-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR