3-(Morpholino)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Morpholino)phenylboronic acid Pinacol ester, 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 2-[3-(Morpholino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8608-5G PN-8608-1G
COBBPN-8608-5GEA 449 EUR
COBBPN-8608-5G COBBPN-8608-1G
3-(Morpholino)phenylboronsäurepinakolester
3-(Morpholino)phenylboronsäurepinakolester
Formel: C₁₆H₂₄BNO₃
Molecular Weight: 289,1777 g/mol
MDL: MFCD03412097
CAS-Nummer: 852227-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR