4-(Morpholino)phenylboronsäurepinakolester
Lieferant: Thermo Scientific
Synonyme:
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine
CC17439.250
CC17439.1
MAYBCC17439.250EA
0
EUR
MAYBCC17439.250
MAYBCC17439.1
4-(Morpholino)phenylboronsäurepinakolester
4-(Morpholino)phenylboronsäurepinakolester
Formel:
C₁₆H₂₄BNO₃ Molecular Weight: 289.18 g/mol |
MDL:
MFCD04112544 CAS-Nummer: 568577-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...