2-(Morpholino)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Morpholinophenyl)boronic acid

BB-8381-1G BB-8381-250M
COBBBB-8381-1GEA 507 EUR
COBBBB-8381-1G COBBBB-8381-250M
2-(Morpholino)phenylboronsäure
2-(Morpholino)phenylboronsäure
Formel: C₁₀H₁₄BNO₃
Molecular Weight: 207,0341 g/mol
MDL: MFCD03095109
CAS-Nummer: 933052-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR