4-(Morpholino)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Morpholinophenyl)boronic acid

BB-8391-5G BB-8391-1G
COBBBB-8391-5GEA 274 EUR
COBBBB-8391-5G COBBBB-8391-1G
4-(Morpholino)phenylboronsäure
4-(Morpholino)phenylboronsäure
Formel: C₁₀H₁₄BNO₃
Molecular Weight: 207,0341 g/mol
MDL: MFCD03095169
CAS-Nummer: 186498-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR