2'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4852-5G CA-4852-1G
COBBCA-4852-5GEA 3080 EUR
COBBCA-4852-5G COBBCA-4852-1G
2'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure
2'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure
Formel: C₁₆H₁₄O₄
MDL: MFCD06210041
CAS-Nummer: 400750-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR