3'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4853-5G CA-4853-1G
COBBCA-4853-5GEA 3080 EUR
COBBCA-4853-5G COBBCA-4853-1G
3'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure
3'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure
Formel: C₁₆H₁₄O₄
MDL: MFCD06210095
CAS-Nummer: 400750-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR