4'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4851-1G CA-4851-5G
COBBCA-4851-1GEA 873 EUR
COBBCA-4851-1G COBBCA-4851-5G
4'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure
4'-(1,3-Dioxolan-2-yl)biphenyl-3-carbonsäure
Formel: C₁₆H₁₄O₄
MDL: MFCD06210080
CAS-Nummer: 400744-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR