5,6-Dimethylbenzotriazol Monohydrat 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5,6-Dimethyl-1,2,3-benzotriazole Hydrate, 5,6-Dimethyl-1H-benzotriazole hydrate, 5,6-Dimethyl-1H-benzotriazole monohydrate, 5,6-Dimethylazimidobenzene monohydrate, 5,6-Dimethyl-1H-benzo[d][1,2,3]triazole hydrate
116050250
ACRO116050250EA
188
EUR
ACRO116050250
5,6-Dimethylbenzotriazol Monohydrat 99%
5,6-Dimethylbenzotriazol Monohydrat
Formel:
C₈H₉N₃·1H₂O Molecular Weight: 165,20 g/mol (147,18 anhy) Schmelzpunkt: 153…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149344 CAS-Nummer: 1354973-50-4 EINECS: 224-058-3 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 150 to 156 °C |
Water | ≤11 % (K.F.) |
Titration with TBAH | ≥98.5 % (on dry substance) |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.003 % |
Substance darkened by H2SO4 | passes test |
Titrable acid | ≤0.0009 meq/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...