5-Chlor-1-(4-piperidinyl)-2-benzimidazolidinon

Lieferant: CHEMPUR

Synonyme: 5-Chloro-1-(piperidin-4-yl)-1H-benzo[d]imidazol-2(3H)-one, 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone

30-0011.5 30-0011.25
CHMP30-0011.5EA 0 EUR
CHMP30-0011.5 CHMP30-0011.25
5-Chlor-1-(4-piperidinyl)-2-benzimidazolidinon
5-Chlor-1-(4-piperidinyl)-2-benzimidazolidinon
Formel: C₁₂H₁₄ClN₃O
Molecular Weight: 251,72 g/mol
MDL: MFCD02093793
CAS-Nummer: 53786-28-0
EINECS: 258-771-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR