Phenethylbiguanid Hydrochlorid, Supelco®
Lieferant: Merck
Synonyme:
Phenformin hydrochloride, 1-Carbamimidamido-N-(2-phenylethyl)methanimidamide Hydrochloride
P7045-10G
P7045-1G
P7045-10GEA
91.4
EUR
P7045-10G
P7045-1G
Phenethylbiguanid Hydrochlorid, Supelco®
Phenethylbiguanid Hydrochlorid
Formel:
C₁₀H₁₆ClN₅ Molecular Weight: 241,7205 g/mol |
MDL:
MFCD00035039 CAS-Nummer: 834-28-6 EINECS: 212-637-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...