1,1-Dimethylbiguanid Hydrochlorid
Lieferant: Cayman Chemical
Synonyme: AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride
13118-1 13118-5
CAYM13118-1EA 31 EUR
CAYM13118-1 CAYM13118-5
1,1-Dimethylbiguanid Hydrochlorid
1,1-Dimethylbiguanid Hydrochlorid
Formel: C₄H₁₂ClN₅
Molecular Weight: 165.625 g/mol
MDL: MFCD00012582
CAS-Nummer: 1115-70-4
EINECS: 214-230-6