1-(4-Chlorphenyl)biguanid Hydrochlorid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B20311.03 B20311.06
B20311.03EA 27.4 EUR
B20311.03 B20311.06
1-(4-Chlorphenyl)biguanid Hydrochlorid ≥97%
1-(4-Chlorphenyl)biguanid Hydrochlorid

Purity: 97% CAS No.: 4022-81-5 Molecular Formula: C8H10ClN5.HCl Formula Weight: 248.12 Melting Point: 247-250° Beilstein Reference: 3732533

Formel: C₈H₁₁Cl₂N₅
Molecular Weight: 248.12 g/mol
Schmelzpunkt: 247…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053020
CAS-Nummer: 4022-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR