1-(4-Fluorphenyl)biguanid Hydrochlorid

Lieferant: FLUOROCHEM

008352-1G 008352-5G
FLUO008352-1GEA 39.9 EUR
FLUO008352-1G FLUO008352-5G
1-(4-Fluorphenyl)biguanid Hydrochlorid
1-(4-Fluorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₁ClFN₅
Molecular Weight: 231,66 g/mol
MDL: MFCD00051986
CAS-Nummer: 16018-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR