Viktoriablau B, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Basic blue 26, N-(4-((4-(Dimethylamino)phenyl)(4-(phenylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride
94890-100G
94890-25G
SIAL94890-100GEA
564
EUR
SIAL94890-100G
SIAL94890-25G
Viktoriablau B, Sigma-Aldrich®
Viktoriablau B
Formel:
C₃₃H₃₂ClN₃ Molecular Weight: 506,803 g/mol |
MDL:
MFCD00011878 CAS-Nummer: 2580-56-5 EINECS: 219-943-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...