5-Nitro-6-(pyrrolidin-1-yl)chinolin

Lieferant: COMBI-BLOCKS

HC-2426-5G HC-2426-1G
COBBHC-2426-5GEA 451 EUR
COBBHC-2426-5G COBBHC-2426-1G
5-Nitro-6-(pyrrolidin-1-yl)chinolin
5-Nitro-6-(pyrrolidin-1-yl)chinolin
Formel: C₁₃H₁₃N₃O₂
MDL: MFCD12026015
CAS-Nummer: 19979-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR