5-Nitro-8-(pyrrolidin-1-yl)chinolin
Lieferant: COMBI-BLOCKS
HC-2353-5G
HC-2353-1G
COBBHC-2353-5GEA
692
EUR
COBBHC-2353-5G
COBBHC-2353-1G
5-Nitro-8-(pyrrolidin-1-yl)chinolin
5-Nitro-8-(pyrrolidin-1-yl)chinolin
Formel:
C₁₃H₁₃N₃O₂ |
MDL:
MFCD00440055 CAS-Nummer: 294194-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...