5,5'-Bis(tributylstannyl)-2,2'-bithiophen, Sigma-Aldrich®

Lieferant: Merck
Gefahr

717614-1G 717614-5G
SIAL717614-1GEA 119 EUR
SIAL717614-1G SIAL717614-5G
5,5'-Bis(tributylstannyl)-2,2'-bithiophen, Sigma-Aldrich®
5,5'-Bis(tributylstannyl)-2,2'-bithiophen
Formel: C₃₂H₅₈S₂Sn₂
MDL: MFCD01319063
CAS-Nummer: 171290-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR