2,5-Bis(tributylstannyl)thiophen, Sigma-Aldrich®

Lieferant: Merck
Gefahr

717703-1G 717703-5G
SIAL717703-1GEA 71.2 EUR
SIAL717703-1G SIAL717703-5G
2,5-Bis(tributylstannyl)thiophen, Sigma-Aldrich®
2,5-Bis(tributylstannyl)thiophen
Formel: C₂₈H₅₆SSn₂
MDL: MFCD13193232
CAS-Nummer: 145483-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR