2-(Tributylstannyl)thiophen

Lieferant: CHEMPUR
Gefahr

KM09996.250 KM09996.1
CHMPKM09996.250EA 0 EUR
CHMPKM09996.250 MAYBKM09996.1
2-(Tributylstannyl)thiophen
2-(Tributylstannyl)thiophen
Formel: C₁₆H₃₀SSn
Molecular Weight: 373,16 g/mol
MDL: MFCD00192513
CAS-Nummer: 54663-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR