2-(Tributylstannyl)thiophen, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
414492-10ML
414492-50ML
SIAL414492-10MLEA
71.8
EUR
SIAL414492-10ML
SIAL414492-50ML
2-(Tributylstannyl)thiophen, Sigma-Aldrich®
2-(Tributylstannyl)thiophen
Formel:
C₁₆H₃₀SSn Molecular Weight: 373,16 g/mol |
MDL:
MFCD00192513 CAS-Nummer: 54663-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...