2-(2-Naphthyl)indol ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-(Naphthalen-2-yl)-1H-indole
B20434.03
B20434.06
B20434.03EA
84.8
EUR
B20434.03
B20434.06
2-(2-Naphthyl)indol ≥98%
2-(2-Naphthyl)indole
Purity: 98%
CAS No.: 23746-81-8
Molecular Formula: C18H13N
Formula Weight: 243.31
MDL No.: MFCD00124144
Formel:
C₁₈H₁₃N Molecular Weight: 243.31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00124144 CAS-Nummer: 23746-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...