6-Brom-1-(tetrahydropyran-2-yl)naphthalin-2-ol

Lieferant: COMBI-BLOCKS

OT-0836-5G OT-0836-1G
COBBOT-0836-5GEA 0 EUR
COBBOT-0836-5G COBBOT-0836-1G
6-Brom-1-(tetrahydropyran-2-yl)naphthalin-2-ol
6-Brom-1-(tetrahydropyran-2-yl)naphthalin-2-ol
Formel: C₁₅H₁₅BrO₂
MDL: MFCD11855958
CAS-Nummer: 1133116-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR